MilliporeSigma
  • Ablation of mouse phosphomannose isomerase (Mpi) causes mannose 6-phosphate accumulation, toxicity, and embryonic lethality.
Ablation of mouse phosphomannose isomerase (Mpi) causes mannose 6-phosphate accumulation, toxicity, and embryonic lethality.
The Journal of biological chemistry (2005-12-13)
Charles DeRossi, Lars Bode, Erik A Eklund, Fangrong Zhang, Joseph A Davis, Vibeke Westphal, Ling Wang, Alexander D Borowsky, Hudson H Freeze
ABSTRACT
MPI encodes phosphomannose isomerase, which interconverts fructose 6-phosphate and mannose 6-phosphate (Man-6-P), used for glycoconjugate biosynthesis. MPI mutations in humans impair protein glycosylation causing congenital disorder of glycosylation Ib (CDG-Ib), but oral mannose supplements normalize glycosylation. To establish a mannose-responsive mouse model for CDG-Ib, we ablated Mpi and provided dams with mannose to rescue the anticipated defective glycosylation. Surprisingly, although glycosylation was normal, Mpi(-/-) embryos died around E11.5. Mannose supplementation even hastened their death, suggesting that man-nose was toxic. Mpi(-/-) embryos showed growth retardation and placental hyperplasia. More than 90% of Mpi(-/-) embryos failed to form yolk sac vasculature, and 35% failed chorioallantoic fusion. We generated primary embryonic fibroblasts to investigate the mechanisms leading to embryonic lethality and found that mannose caused a concentration- and time-dependent accumulation of Man 6-P in Mpi(-/-) fibroblasts. In parallel, ATP decreased by more than 70% after 24 h compared with Mpi(+/+) controls. In cell lysates, Man-6-P inhibited hexokinase (70%), phosphoglucose isomerase (65%), and glucose-6-phosphate dehydrogenase (85%), but not phosphofructokinase. Incubating intact Mpi(-/-) fibroblasts with 2-[(3)H]deoxyglucose confirmed mannose-dependent hexokinase inhibition. Our results in vitro suggest that mannose toxicity in Mpi(-/-) embryos is caused by Man-6-P accumulation, which inhibits glucose metabolism and depletes intracellular ATP. This was confirmed in E10.5 Mpi(-/-) embryos where Man-6-P increased more than 10 times, and ATP decreased by 50% compared with Mpi(+/+) littermates. Because Mpi ablation is embryonic lethal, a murine CDG-Ib model will require hypomorphic Mpi alleles.
Product Description
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood