MilliporeSigma
  • Hyaluronic acid: additional biochemical marker in the diagnosis of biliary atresia.
Hyaluronic acid: additional biochemical marker in the diagnosis of biliary atresia.
Pediatrics international : official journal of the Japan Pediatric Society (2007-09-19)
Nuthapong Ukarapol, Lumduan Wongsawasdi, Siriwan Ong-Chai, Pornthawee Riddhiputra, Prachya Kongtawelert
ABSTRACT
The purpose of the present paper was to evaluate the value of biochemical markers, including conventional liver function tests, gamma-glutamyl transferase (GGT), and hyaluronic acid (HA), in the diagnosis of neonatal cholestasis. Infants with neonatal jaundice were consecutively enrolled during 1 year period. The patients were diagnosed as having biliary atresia (BA) if there was either bile ductular proliferation in the portal tracts, atretic common bile duct/gallbladder, or evidence of bile duct obstruction demonstrated by liver pathology or intraoperative cholangiography, respectively. Serum HA was measured using an enzyme-linked immunosorbent assay-based test. A total of 25 patients diagnosed as having BA (n = 10), neonatal hepatitis (NH; n = 9), choledochal cyst (n = 3) and parenteral nutrition-induced cholestasis (n = 3), were studied. The age at diagnosis was not significantly different between groups. Only GGT and HA were significantly elevated in the patients with BA when compared to NH (P = 0.02, P = 0.03, respectively). In BA, the median value of serum HA was 514 ng/mL (range 19-4476 ng/mL), compared to 50 ng/mL (range 19-315 ng/mL) in NH. Additionally, the serum HA level was much higher in children with choledochal cyst. HA could be considered as a complementary biochemical marker for evaluating infants with prolonged jaundice.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Chenodeoxycholic acid
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$236.00
$165.20