MilliporeSigma
  • Effect of alpha-1-acid glycoprotein, albumin and palmitic acid on the brain and salivary gland extraction of warfarin in rats.
Effect of alpha-1-acid glycoprotein, albumin and palmitic acid on the brain and salivary gland extraction of warfarin in rats.
The Journal of pharmacology and experimental therapeutics (1989-02-01)
S Urien, D Morin, J P Tillement
ABSTRACT
The effect of plasma protein binding of warfarin on its transfer into the brain and salivary gland was investigated using alpha-1-acid glycoprotein and human serum albumin (HSA) in combination or not with palmitic acid. The tissue extraction of [14C] warfarin relative to [3H]water was determined by intracarotid injection technique in male Wistar rats. The tissue extraction of warfarin varied inversely with the concentration of added serum protein, (HSA and alpha-1-acid glycoprotein), and addition of palmitic acid to HSA diminished the extraction. The fraction of drug uptaked by tissue (tissue available fraction) was always dramatically greater than the in vitro free drug fraction, and this was interpreted as an enhanced in vivo drug dissociation from the binding protein. The fraction of drug uptake by salivary gland was closer to the in vitro free fraction than the fraction of drug uptake by brain tissue. The addition of palmitic acid to HSA induced parallel changes in the in vitro free fraction of warfarin and in the brain tissue or salivary gland extraction of warfarin. These data indicate that a part of protein-bound warfarin (as determined in vitro) is available for tissue extraction via an enhanced in vivo dissociation of the drug-protein complex in the tissue microcirculation. The in vitro data were fitted to a saturable model of binding whereas the in vivo data could satisfactorily fit a model dealing with a nonsaturable model of binding, and this is probably the result of the several-fold increase in the in vivo dissociation constant.
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Sigma-Aldrich
Palmitic acid, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Supelco
Palmitic acid, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$135.00
25 mg
In Stock
Details...
$523.00