MilliporeSigma
  • Determination of nicotine and four metabolites in the serum of smokers by high-performance liquid chromatography with ultraviolet detection.
Determination of nicotine and four metabolites in the serum of smokers by high-performance liquid chromatography with ultraviolet detection.
Journal of chromatography (1993-11-24)
P Zuccaro, I Altieri, M Rosa, A R Passa, S Pichini, G Ricciarello, R Pacifici
ABSTRACT
A simple and reliable reversed-phase high-performance liquid chromatographic method with ultraviolet detection is described for the quantitation of nicotine and its metabolites cotinine, trans-3'-hydroxycotinine, norcotinine and cotinine N-oxide in human serum. The analytes and the internal standard, N-ethylnorcotinine, were extracted by solid-phase extraction before chromatography. Two different columns and mobile phases with gradient systems were used. The detection limit of the assay was 10 ng/ml for nicotine, 3 ng/ml for cotinine N-oxide and 5 ng/ml for cotinine, trans-3'-hydroxycotinine and norcotinine. The concentrations of nicotine and its metabolites in the serum of 12 cigarette smokers are reported.
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Sigma-Aldrich
4-Methylcatechol, ≥95%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Supelco
S(−)-Nicotine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®