MilliporeSigma
  • Novel antagonists of alcohol inhibition of l1-mediated cell adhesion: multiple mechanisms of action.
Novel antagonists of alcohol inhibition of l1-mediated cell adhesion: multiple mechanisms of action.
Molecular pharmacology (2002-10-23)
Michael F Wilkemeyer, Carrie E Menkari, Michael E Charness
ABSTRACT
1-Octanol antagonizes ethanol inhibition of L1-mediated cell adhesion and prevents ethanol teratogenesis in mouse whole embryo culture. Herein, we identify a new series of alcohol antagonists and study their mechanism of action. Cell aggregation assays were carried out in ethanol-sensitive, human L1-transfected NIH/3T3 cells in the absence and presence of 100 mM ethanol or 2 mM 1-butanol and candidate antagonists. Antagonist potency for 1-alcohols increased progressively over 5 log orders from 1-pentanol (C5) to 1-dodecanol (C12). Antagonist potency declined from 1-dodecanol (C12) to 1-tridecanol (C13), and 1-tetradecanol (C14) and 1-pentadecanol (C15) were inactive. The presence and position of a double bond in the 1-butanol molecule determined whether a compound was a full agonist (1-butanol), a mixed agonist-antagonist (2-buten-1-ol), or an antagonist (3-buten-1-ol). Increasing the concentration of agonist (1-butanol or ethanol) overcame the antagonism of 3-buten-1-ol, benzyl alcohol, cyclopentanol, and 3-pentanol, but not that of 4-methyl-1-pentanol, 2-methyl-2-pentanol, 1-pentanol, 2-pentanol, 1-octanol, and 2,6-di-isopropylphenol (propofol), suggesting that the mechanisms of antagonism may differ between these groups of compounds. These findings suggest that selective straight, branched, and cyclic alcohols may act at multiple, discrete sites to antagonize the actions of ethanol and 1-butanol on L1-mediated cell-cell adhesion.
Sigma-Aldrich
2-Methyl-2-pentanol, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on February 07, 2025
Details...
₩118,451