MilliporeSigma
  • The art of "cut and run": the role of Rab14 GTPase in regulating N-cadherin shedding and cell motility.
The art of "cut and run": the role of Rab14 GTPase in regulating N-cadherin shedding and cell motility.
Developmental cell (2012-05-19)
Rytis Prekeris
ABSTRACT
Linford et al. define a Rab14-mediated endocytic recycling pathway that controls proteolytic N-cadherin cleavage by transporting ADAM10 protease to the plasma membrane. When this pathway is disrupted, diminished ADAM10-dependent N-cadherin shedding leads to increased cell-cell adhesion and inhibition of cell motility.