MilliporeSigma
  • Economical Pt-free catalysts for counter electrodes of dye-sensitized solar cells.
Economical Pt-free catalysts for counter electrodes of dye-sensitized solar cells.
Journal of the American Chemical Society (2012-01-28)
Mingxing Wu, Xiao Lin, Yudi Wang, Liang Wang, Wei Guo, Daidi Qi, Xiaojun Peng, Anders Hagfeldt, Michael Grätzel, Tingli Ma
ABSTRACT
Three classes (carbides, nitrides and oxides) of nanoscaled early-transition-metal catalysts have been proposed to replace the expensive Pt catalyst as counter electrodes (CEs) in dye-sensitized solar cells (DSCs). Of these catalysts, Cr(3)C(2), CrN, VC(N), VN, TiC, TiC(N), TiN, and V(2)O(3) all showed excellent catalytic activity for the reduction of I(3)(-) to I(-) in the electrolyte. Further, VC embedded in mesoporous carbon (VC-MC) was prepared through in situ synthesis. The I(3)(-)/I(-) DSC based on the VC-MC CE reached a high power conversion efficiency (PCE) of 7.63%, comparable to the photovoltaic performance of the DSC using a Pt CE (7.50%). In addition, the carbide catalysts demonstrated catalytic activity higher than that of Pt for the regeneration of a new organic redox couple of T(2)/T(-). The T(2)/T(-) DSCs using TiC and VC-MC CEs showed PCEs of 4.96 and 5.15%, much higher than that of the DSC using a Pt CE (3.66%). This work expands the list of potential CE catalysts, which can help reduce the cost of DSCs and thereby encourage their fundamental research and commercial application.
Sigma-Aldrich
Vanadium(IV) carbide, powder, <2 μm, 99.9% trace metals basis