MilliporeSigma
  • New advances in the understanding of the in-source decay fragmentation of peptides in MALDI-TOF-MS.
New advances in the understanding of the in-source decay fragmentation of peptides in MALDI-TOF-MS.
Journal of the American Society for Mass Spectrometry (2010-09-14)
Kevin Demeure, Valérie Gabelica, Edwin Andre De Pauw
ABSTRACT
In-source decay (ISD) is a rapid fragmentation occurring in the matrix-assisted laser desorption/ionization (MALDI) source before the ion extraction. Despite the increasing interest for peptides de novo sequencing by ISD, the influence of the matrix and of the peptide itself is not yet fully understood. Here we compare matrices with high ISD efficiencies to gain deeper insight in the ISD fragmentation process(es). The major ISD fragments are the c- and z-ions, but other types of fragments are also observed, and their origin is studied here. Two main pathways lead to fragmentation in the source: a radical-induced pathway that leads to c-, z-, w-, and d-ions, and a thermally activated pathway that leads to y-, b-, and a-ions. A detailed analysis of the ISD spectra of selected peptides revealed that (1) the extents of the two in-source pathways are differently favored depending on the matrix used, that (2) the presence of a positive/negative charge on the radical-induced fragments is necessary for their observation in positive/negative mode, respectively, and that (3), for a same peptide, the patterns of the different types of fragments differ according to the matrix used.
Product Description
Supelco
1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)