MilliporeSigma
Evaluation of benzoic acid derivatives as sirtuin inhibitors.
Bioorganic & medicinal chemistry letters (2013-11-26)
Yi-Pei Chen, Chad C Catbagan, Jeannette T Bowler, Trevor Gokey, Natalie D M Goodwin, Anton B Guliaev, Weiming Wu, Taro Amagata
ABSTRACT
Employing a genetically modified yeast strain as a screening tool, 4-dimethylaminobenzoic acid (5) was isolated from the marine sediment-derived Streptomyces sp. CP27-53 as a weak yeast sirtuin (Sir2p) inhibitor. Using this compound as a scaffold, a series of disubstituted benzene derivatives were evaluated to elucidate the structure activity relationships for Sir2p inhibition. The results suggested that 4-alkyl or 4-alkylaminobenzoic acid is the key structure motif for Sir2p inhibitory activity. The most potent Sir2p inhibitor, 4-tert-butylbenzoic acid (20), among the tested compounds in this study turned out to be a weak but selective SIRT1 inhibitor. The calculated binding free energies between the selected compounds and the catalytic domain of SIRT1 were well correlated to their measured SIRT1 inhibitory activities.
Sigma-Aldrich
4-tert-Butylbenzoic acid, 99%