MilliporeSigma
  • gfp-based N-acyl homoserine-lactone sensor systems for detection of bacterial communication.
gfp-based N-acyl homoserine-lactone sensor systems for detection of bacterial communication.
Applied and environmental microbiology (2001-02-07)
J B Andersen, A Heydorn, M Hentzer, L Eberl, O Geisenberger, B B Christensen, S Molin, M Givskov
ABSTRACT
In order to perform single-cell analysis and online studies of N-acyl homoserine lactone (AHL)-mediated communication among bacteria, components of the Vibrio fischeri quorum sensor encoded by luxR-P(luxI) have been fused to modified versions of gfpmut3* genes encoding unstable green fluorescent proteins. Bacterial strains harboring this green fluorescent sensor detected a broad spectrum of AHL molecules and were capable of sensing the presence of 5 nM N-3-oxohexanoyl-L-homoserine lactone in the surroundings. In combination with epifluorescent microscopy, the sensitivity of the sensor enabled AHL detection at the single-cell level and allowed for real-time measurements of fluctuations in AHL concentrations. This green fluorescent AHL sensor provides a state-of-the-art tool for studies of communication between the individuals present in mixed bacterial communities.
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
$70.80