MilliporeSigma
  • Beta-galactosidase catalyzed selective galactosylation of aromatic compounds.
Beta-galactosidase catalyzed selective galactosylation of aromatic compounds.
Biotechnology progress (2006-02-04)
Nicolas Bridiau, Selma Taboubi, Nejib Marzouki, Marie Dominique Legoy, Thierry Maugard
ABSTRACT
A new approach to galacto-oligosaccharides and galacto-conjugates synthesis performed by the beta-galactosidase from Kluyveromyces lactis is reported. The enzymatic galactosylation of eight kinds of adsorbed aromatic primary alcohols, in particular the two drugs guaifenesin and chlorphenesin, gave the corresponding beta-D-galacto-pyranosides in yields ranging between approximately 10% and 96%. For the first time, we have showed that the adsorption of acceptor substrates onto solid supports such as silica gel influences the yield and the selectivity of galacto-conjugates synthesis. In particular, we observed that adsorption of acceptor favored the synthesis of digalactosylated compounds.
Sigma-Aldrich
3-Aminobenzyl alcohol, 97%