MilliporeSigma
  • Porcine pancreatic lipase related protein 2 has high triglyceride lipase activity in the absence of colipase.
Porcine pancreatic lipase related protein 2 has high triglyceride lipase activity in the absence of colipase.
Biochimica et biophysica acta (2013-06-19)
Xunjun Xiao, Leah E Ross, Wednesday A Sevilla, Yan Wang, Mark E Lowe
ABSTRACT
Efficient dietary fat digestion is essential for newborns who consume more dietary fat per body weight than at any other time of life. In many mammalian newborns, pancreatic lipase related protein 2 (PLRP2) is the predominant duodenal lipase. Pigs may be an exception since PLRP2 expression has been documented in the intestine but not in the pancreas. Because of the differences in tissue-specific expression, we hypothesized that the kinetic properties of porcine PLRP2 would differ from those of other mammals. To characterize its properties, recombinant porcine PLRP2 was expressed in HEK293T cells and purified to homogeneity. Porcine PLRP2 had activity against tributyrin, trioctanoin and triolein. The activity was not inhibited by bile salts and colipase, which is required for the activity of pancreatic triglyceride lipase (PTL), minimally stimulated PLRP2 activity. Similar to PLRP2 from other species, PLRP2 from pigs had activity against galactolipids and phospholipids. Importantly, porcine PLRP2 hydrolyzed a variety of dietary substrates including pasteurized human mother's milk and infant formula and its activity was comparable to that of PTL. In conclusion, porcine PLRP2 has broad substrate specificity and has high triglyceride lipase activity even in the absence of colipase. The data suggest that porcine PLRP2 would be a suitable lipase for inclusion in recombinant preparations for pancreatic enzyme replacement therapy.
Sigma-Aldrich
Glyceryl trioleate, ≥97.0% (TLC)
Sigma-Aldrich
Glyceryl tributyrate, puriss., ≥98.5% (GC)
Sigma-Aldrich
Glyceryl trioctanoate, ≥95% (GC)
Sigma-Aldrich
Glyceryl trioleate, ≥99%
Sigma-Aldrich
Glyceryl trioleate, ~65%
Sigma-Aldrich
Glyceryl tributyrate, ≥99%