MilliporeSigma
  • Prediction of fatty acids content in pig adipose tissue by near infrared spectroscopy: at-line versus in-situ analysis.
Prediction of fatty acids content in pig adipose tissue by near infrared spectroscopy: at-line versus in-situ analysis.
Meat science (2013-06-25)
E Zamora-Rojas, A Garrido-Varo, E De Pedro-Sanz, J E Guerrero-Ginel, D Pérez-Marín
ABSTRACT
A handheld micro-electro-mechanical system (MEMS) based spectrometer working in the near infrared region (NIR) (1600-2400nm) was evaluated for in-situ and non-destructive prediction of main fatty acids in Iberian pig (IP) carcasses. 110 IP carcasses were measured. Performance of the instrument was compared with at-line high-resolution NIRS monochromators working in two analysis modes: melted fat samples (transflectance cups) and intact adipose tissues (interactance fiber optic). Standard Error of Prediction (SEP) values obtained on the MEMS-NIRS device were: 0.68% (stearic), 1.30% (oleic), 0.55% (linoleic) and 1% (palmitic), explaining a variability of 83%, 84%, 81% and 78%, respectively. As expected, this represented a loss of predictive capability in comparison to at-line models, even with the same spectral characteristics as on the handheld device. However, the estimated total errors were at the same level for gas chromatography and NIRS analysis. This indicates that the MEMS-NIRS in-situ analysis of each individual carcass provides a cost-effective and real-time quality control system with suitable accuracy.
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linoleic acid, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palmitic acid, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Palmitic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Linoleic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
SKU
Pack Size
Availability
Price
Quantity
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Stearic acid, reagent grade, 95%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Oleic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Stearic acid, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, technical grade, 90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, natural, FCC
SKU
Pack Size
Availability
Price
Quantity
Stearic acid, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oleic acid, SAJ first grade, ≥70.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Supelco
Oleic acid, Selectophore, ≥99%