MilliporeSigma
In vitro efficacy of ocular surface lubricants against dehydration.
Cornea (2013-07-19)
Xiaodong Zheng, Tomoko Goto, Atsushi Shiraishi, Yuichi Ohashi
ABSTRACT
To compare 3 ocular lubricants containing sodium hyaluronate (SH), carboxymethylcellulose (CMC), and hydroxypropyl methylcellulose (HPMC) for their ability to enhance water retention and to protect human corneal epithelial cells (HCECs) from dehydration. Experiments were performed using 0.1% and 0.3% solutions of the 3 lubricants diluted in Milli-Q water for the water retention assays and in DMEM/F12 culture medium for the cell viability assays. Five milliliters of each of the lubricants was dropped onto a filter paper, and the paper was kept in an open container at 25°C and a humidity of 36% to 38%. The weight of the paper was measured hourly for 4 hours. In the second set of experiment, cultured HCECs were exposed to the test lubricants for 60 minutes, and the lubricants were removed. Cells were then exposed to room air for up to 60 minutes. Cells were then incubated with a vital dye, and the absorption of the reduced form of the dye was measured. The cell survival rate was compared for the 3 lubricants. Filter papers moistened with both 0.1% and 0.3% SH were significantly heavier than those moistened with CMC and HPMC at all time points. The survival rate of HCECs was significantly higher at most times with 0.1% and 0.3% SH than with CMC and HPMC solutions. The effects of CMC were not significantly different from those of HPMC. These findings indicate that SH is significantly better for water retention and protection of HCECs from dehydration than HPMC and CMC.
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Carboxymethylcellulose sodium salt, SAJ first grade
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity