MilliporeSigma
  • A modified ninhydrin micro-assay for determination of total cyanogens in plants.
A modified ninhydrin micro-assay for determination of total cyanogens in plants.
Food chemistry (2013-07-23)
Andriana Surleva, Gabi Drochioiu
ABSTRACT
Indirect quantification of total cyanogenic compounds (cyanogens) in plants was studied using a novel ninhydrin-based spectrophotometric micromethod. The ninhydrin-cyanide system obeys Beer's law in the range from 20 μg L(-1) to 800 μg L(-1) CN(-) with molar absorptivity ε=1.4×10(5) L mol(-1)cm(-1). Recovery at 100 μg L(-1) CN(-) was 100.5±0.1% and LOD and LOQ were 8 and 22 μg L(-1), respectively. The conditions for ninhydrin reagent preparation were discussed. The extraction and separation solutions, extraction and incubation time, and solvent volume were also investigated for maximum recovery of total cyanogens. 0.1% NaHCO3 was used as an extraction solvent for cyanide formed after enzymatic hydrolysis of cyanogens. The procedure was suitable for samples containing more than 90 mg CN(-)/100 g sample. When cyanogen concentrations were lower, the resulted cyanide was separated by microdiffusion in a Conway cell. Water was used as a hydrolysis medium and a donor solvent, while 2% Na2CO3 as an absorbing solution. Total cyanogen content in plum and almond kernels, as well as apple and flax seeds was determined.
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Ninhydrin, SAJ special grade, ≥98.0%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection