MilliporeSigma
  • Sweet anion receptors: recognition of chiral carboxylate anions by D-glucuronic-acid-decorated diindolylmethane.
Sweet anion receptors: recognition of chiral carboxylate anions by D-glucuronic-acid-decorated diindolylmethane.
Organic letters (2013-08-31)
Jarosław M Granda, Janusz Jurczak
ABSTRACT
Anion receptors containing glucuronic acid were synthesized, and their anion binding ability studied. Chirality of anionic guests derived from mandelic acid and amino acids can be distinguished not only in terms of stability constants but also by significant differences in chemical shift changes for sugar moiety protons.
Sigma-Aldrich
(R)-(−)-Mandelic acid, ChiPros®, produced by BASF, 99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, 98%
Sigma-Aldrich
Mandelic acid, 99%
Sertraline impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
DL-Mandelic acid, analytical standard