MilliporeSigma
Oral syphilis: a series of 5 cases.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2013-09-21)
Moritz Hertel, Daniel Matter, Andrea M Schmidt-Westhausen, Michael M Bornstein
ABSTRACT
Syphilis is an infectious, usually sexually transmitted, disease caused by Treponema pallidum, subspecies pallidum. Because of the increasing prevalence in Europe during the past few years, dentists could be confronted with patients with oral manifestations of syphilis. Because oral lesions are highly contagious, it is vital to make the correct diagnosis quickly to initiate the proper therapy and to interrupt the chain of infection. We present the cases of 5 patients with syphilis-related oral lesions. These cases are representative because of their clinical presentation, age, and gender distribution and the diagnostic approach. The aim of the present report is to emphasize the importance of the dentist knowing and identifying syphilis in different stages to diagnose the disease and institute treatment at an early stage.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Penicillin G potassium salt
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard