MilliporeSigma
  • Stress resistance of Escherichia coli and Bacillus subtilis is modulated by auxins.
Stress resistance of Escherichia coli and Bacillus subtilis is modulated by auxins.
Canadian journal of microbiology (2013-11-12)
J Repar, S Šućurović, K Zahradka, D Zahradka, M Ćurković-Perica
ABSTRACT
Two bacterial species, Gram-negative Escherichia coli and Gram-positive Bacillus subtilis, were exposed to different auxins to examine possible effects of these substances on bacterial stress tolerance. Bacterial resistance to UV irradiation, heat shock, and streptomycin was assessed with and without previous exposure to the following auxins: indole-3-acetic acid (IAA), indole-3-butyric acid (IBA), and 1-naphthalene acetic acid (NAA). Escherichia coli and B. subtilis cultures pretreated with any of the 3 auxins survived UV irradiation better than the untreated cultures. Also, B. subtilis cultures pretreated with IBA or NAA survived prolonged heat exposure better than the untreated cultures, while IAA pretreatment had no effect on heat shock survival. In contrast, auxin pretreatment rendered E. coli more sensitive to heat shock. Escherichia coli cultures pretreated with auxins were also more sensitive to streptomycin, while auxin pretreatment had no effect on sensitivity of B. subtilis to streptomycin. These results show that auxins may either enhance or reduce bacterial tolerance to different stressors, depending on the bacterial species and the type and level of the stress. Auxins usually had similar effects on the same bacterial species in cases when the same type and level of stress were applied.
Product Description
Supelco
1-Naphthaleneacetic acid, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
3-Indoleacetic acid, SAJ special grade, ≥98.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Supelco
Streptomycin sesquisulfate hydrate, VETRANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Indole-3-butyric acid, BioReagent, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Indole-3-butyric acid, ≥98.0% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
1-Naphthylacetic acid, BioReagent, 1 mg/mL, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on January 22, 2025
$93.40
5 g
Estimated to ship on January 22, 2025
$201.00
25 g
Estimated to ship on January 22, 2025
$713.00