- The dizzy child.
ABSTRACT
Dizziness in childhood may be the result of significant vestibular or central nervous system pathology. It is the responsibility of otolaryngologists and neurologists to provide appropriate diagnosis and treatment of these disorders.