MilliporeSigma
  • Highly sensitive and selective Pd2+ sensor of naphthalimide derivative based on complexation with alkynes and thio-heterocycle.
Highly sensitive and selective Pd2+ sensor of naphthalimide derivative based on complexation with alkynes and thio-heterocycle.
Chemical communications (Cambridge, England) (2008-12-03)
Liping Duan, Yufang Xu, Xuhong Qian
ABSTRACT
A new fluorescent Pd2+ sensor , N-butyl-4-(p-methyloxy)-phenylethynyl-5-thiophenemethylamino-1,8-naphthalimide, was designed and synthesized. It showed highly selective on-off fluorescence changes for Pd2+ among the representative transition and heavy metallic cations, and its fluorescence was efficiently quenched by 5 equivalents of Pd2+ in buffer solution. The sensor also displayed a selective chromogenic behavior toward Pd2+ from yellow to black-red, which could be easily observed by the naked eye.
Sigma-Aldrich
2-Thiophenemethylamine, 96%