MilliporeSigma
  • The neuroendocrine protein 7B2 suppresses the aggregation of neurodegenerative disease-related proteins.
The neuroendocrine protein 7B2 suppresses the aggregation of neurodegenerative disease-related proteins.
The Journal of biological chemistry (2012-11-23)
Michael Helwig, Akina Hoshino, Casey Berridge, Sang-Nam Lee, Nikolai Lorenzen, Daniel E Otzen, Jason L Eriksen, Iris Lindberg
ABSTRACT
Neurodegenerative diseases such as Alzheimer (AD) and Parkinson (PD) are characterized by abnormal aggregation of misfolded β-sheet-rich proteins, including amyloid-β (Aβ)-derived peptides and tau in AD and α-synuclein in PD. Correct folding and assembly of these proteins are controlled by ubiquitously expressed molecular chaperones; however, our understanding of neuron-specific chaperones and their involvement in the pathogenesis of neurodegenerative diseases is limited. We here describe novel chaperone-like functions for the secretory protein 7B2, which is widely expressed in neuronal and endocrine tissues. In in vitro experiments, 7B2 efficiently prevented fibrillation and formation of Aβ(1-42), Aβ(1-40), and α-synuclein aggregates at a molar ratio of 1:10. In cell culture experiments, inclusion of recombinant 7B2, either in the medium of Neuro-2A cells or intracellularly via adenoviral 7B2 overexpression, blocked the neurocytotoxic effect of Aβ(1-42) and significantly increased cell viability. Conversely, knockdown of 7B2 by RNAi increased Aβ(1-42)-induced cytotoxicity. In the brains of APP/PSEN1 mice, a model of AD amyloidosis, immunoreactive 7B2 co-localized with aggregation-prone proteins and their respective aggregates. Furthermore, in the hippocampus and substantia nigra of human AD- and PD-affected brains, 7B2 was highly co-localized with Aβ plaques and α-synuclein deposits, strongly suggesting physiological association. Our data provide insight into novel functions of 7B2 and establish this neural protein as an anti-aggregation chaperone associated with neurodegenerative disease.
Sigma-Aldrich
Anti-Histidine Tagged Antibody, clone 4D11, Alexa Fluor 488 conjugate, clone 4D11, Upstate®, from mouse
Sigma-Aldrich
α-Lactalbumin from bovine milk, Marker for non-denaturing PAGE