MilliporeSigma
  • New insecticidal rocaglamide derivatives and related compounds from Aglaia oligophylla.
New insecticidal rocaglamide derivatives and related compounds from Aglaia oligophylla.
Journal of natural products (2001-04-28)
M Dreyer, B W Nugroho, F I Bohnenstengel, R Ebel, V Wray, L Witte, G Bringmann, J Mühlbacher, M Herold, P D Hung, L C Kiet, P Proksch
ABSTRACT
Organic-soluble extracts of the twigs of Aglaia oligophylla collected in Vietnam yielded four insecticidal cyclopentatetrahydrobenzofurans of the rocaglamide type including one new natural product (compound 4). Moreover, two cyclopentatetrahydrobenzopyran derivatives, belonging to the aglain and aglaforbesin types, respectively, were also isolated. The aglaforbesin derivative 6 proved likewise to be a new natural product. All isolated rocaglamide, aglain, and aglaforbesin derivatives have a characteristic methylenedioxy substituent linked to C-6 and C-7 or to C-7 and C-8, respectively. Structure elucidation of the new natural products and the determination of the absolute configuration of compound 1 by calculation of its CD spectrum with molecular dynamics simulation are described. All isolated rocaglamide derivatives exhibited strong insecticidal activity toward neonate larvae of the polyphageous pest insect Spodoptera littoralis when incorporated into an artificial diet, with LC(50) values varying between 2.15 and 6.52 ppm.
Sigma-Aldrich
Rocaglamide, ≥96% (HPLC)