Merck
Accidental etizolam ingestion in a child.
Pediatric emergency care (2007-08-02)
Zenichiro Kato, Mitsuhiro Nakamura, Michinori Funato, Hideaki Kuwabara, Naomi Kondo
ABSTRACT
Etizolam (ETZ) is an antidepressive thienodiazepine drug that is used worldwide. The most frequent adverse effects in adults are drowsiness and muscle weakness, and this can rarely cause paradoxical excitation; however, no information exists on intoxication in children. Furthermore, evidence bearing on its safety in children is not available. We present a case of a child who accidentally took a single dose of ETZ, approximately the same as a therapeutic dose for adults, and who showed paradoxical excitation and muscle weakness. The case presented here suggests that pediatricians and emergency physicians should be aware of the possible adverse effects in children and therapeutic approaches in intoxication of ETZ and the necessity of further investigations on a specific therapeutic guideline for overdose management especially in children.
Product Description
Supelco
Etizolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®