MilliporeSigma
  • Once-daily administration of trospium chloride extended release provides 24-hr coverage of nocturnal and diurnal symptoms of overactive bladder: an integrated analysis of two phase III trials.
Once-daily administration of trospium chloride extended release provides 24-hr coverage of nocturnal and diurnal symptoms of overactive bladder: an integrated analysis of two phase III trials.
Neurourology and urodynamics (2011-01-27)
David A Ginsberg, Michael G Oefelein, Pamela I Ellsworth
ABSTRACT
Once-daily extended release (XR) trospium chloride, which provides therapeutic trospium plasma concentrations over 24 hours, has demonstrated efficacy in treating overactive bladder (OAB) symptoms as evaluated over a 24-hr period. This analysis examined the effects of trospium XR on diurnal and nocturnal OAB symptoms. Pooled data were analyzed from two identically designed Phase III trials in which patients with OAB were randomized to receive trospium XR 60 mg or placebo once daily in the morning for 12 weeks. Efficacy was assessed using 3-day urinary diaries. Diurnal events were those occurring from arising from bed in the morning until retiring in the evening; nocturnal events were those occurring from retiring until arising. In total, 1,165 patients received trospium XR (N = 578) or placebo (N = 587). At Week 12 comparison of trospium XR versus placebo, a significantly greater mean reduction from baseline in nocturnal voids (-0.8 vs. -0.6; P = 0.006) and diurnal voids (-1.9 vs. -1.4; P < 0.0001) was noted. At Week 12, the mean percent reduction from baseline with trospium XR versus placebo in nocturnal urge urinary incontinence (UUI) episodes (-60.2% vs. -48.3%; P = 0.003) and mean absolute reduction in diurnal UUI episodes (-2.0 vs. -1.5; P < 0.0001) was significantly greater. Predictors of nocturnal response were duration (weeks) and type of therapy (trospium XR vs. placebo). Reductions in nocturnal toilet voids were accompanied by significant improvements in sleep-related quality of life (QoL) domains. Trospium XR significantly improved both nocturnal and diurnal OAB symptoms. Nocturnal improvements were associated with sleep-related QoL benefits. These results indicate that the XR formulation of trospium chloride provides effective 24-hr coverage of OAB symptoms.
Product Description
Trospium chloride, European Pharmacopoeia (EP) Reference Standard