Merck
  • Fixed-combination brinzolamide 1%/brimonidine 0.2% vs monotherapy with brinzolamide or brimonidine in patients with open-angle glaucoma or ocular hypertension: results of a pooled analysis of two phase 3 studies.
Fixed-combination brinzolamide 1%/brimonidine 0.2% vs monotherapy with brinzolamide or brimonidine in patients with open-angle glaucoma or ocular hypertension: results of a pooled analysis of two phase 3 studies.
Eye (London, England) (2013-05-04)
T Realini, Q H Nguyen, G Katz, H DuBiner
ABSTRACT
To describe pooled efficacy and safety data from two phase 3 studies comparing brinzolamide 1%/brimonidine 0.2% fixed combination (BBFC) with its component medications, brinzolamide and brimonidine, in patients with open-angle glaucoma or ocular hypertension. Data were pooled from two nearly identical clinical trials comparing BBFC with its component medications, each given three times daily. The 3-month efficacy outcome was mean intraocular pressure (IOP) at 0800, 1000, 1500, and 1700 hours. Safety outcomes included adverse events (AEs), best-corrected visual acuity, examination of ocular structures, pachymetry, perimetry, and vital signs. A total of 1350 patients were enrolled and included in this analysis (BBFC, n=437; brinzolamide, n=458; brimonidine, n=455). Baseline mean IOP levels were similar among the three treatment groups. At 3 months, mean IOP of the BBFC group was significantly lower than that of either monotherapy group (P<0.0001) at all the four time points. A total of 272 patients (20.1%) experienced at least one treatment-related AE (BBFC, 24.6%; brinzolamide, 18.7%; brimonidine, 17.4%), the majority of which were ocular AEs. One serious AE, moderate intensity chest pain, was considered related to brinzolamide treatment and resulted in study discontinuation. This analysis strengthens the conclusions drawn from the two individual phase 3 studies showing that, in patients with open-angle glaucoma or ocular hypertension, BBFC had significantly superior IOP-lowering activity compared with either brinzolamide or brimonidine alone and a safety profile consistent with that of its individual components.
Product Description
Brimonidine tartrate, European Pharmacopoeia (EP) Reference Standard