MilliporeSigma
  • Lupus erythematosus/lichen planus overlap syndrome: successful treatment with acitretin.
Lupus erythematosus/lichen planus overlap syndrome: successful treatment with acitretin.
Lupus (2013-06-14)
D J Lospinoso, C Fernelius, K D Edhegard, D R Finger, N S Arora
ABSTRACT
Lupus erythematosus/lichen planus overlap syndrome is a rare disorder combining the clinical, histological and immunopathological features of both lupus erythematosus (LE) and lichen planus (LP). Cutaneous lesions mostly affect the distal arms, legs, face and trunk. Palmoplantar involvement is felt to be characteristic of this condition. Plaques are often painful, centrally atrophic, bluish-red to hypopigmented in color, large, and scaly. On biopsy of clinically ambiguous lesions, histopathological features of one or both processes can be found, obscuring the diagnosis and complicating prognosis and treatment. Thus, direct immunofluorescence has become an essential tool in helping to diagnose this condition. In this report we describe the unique clinical and immunohistopathological manifestations of lupus erythematosus/lichen planus overlap syndrome along with a successful response to treatment with acitretin.
Product Description
USP
Acitretin Related Compound A, United States Pharmacopeia (USP) Reference Standard