MilliporeSigma
  • Synthesis, characterization, and antimicrobial activity of poly(acrylonitrile-co-methyl methacrylate) with silver nanoparticles.
Synthesis, characterization, and antimicrobial activity of poly(acrylonitrile-co-methyl methacrylate) with silver nanoparticles.
Applied biochemistry and biotechnology (2013-07-23)
M R El-Aassar, Elsayed E Hafez, Moustafa M G Fouda, Salem S Al-Deyab
ABSTRACT
Nanotechnology is expected to open some new aspects to fight and prevent diseases using atomic-scale tailoring of materials. The main aim of this study is to biosynthesize silver nanoparticles (AgNPs) using Trichoderma viride (HQ438699); the metabolite of this fungus will help either in reduction of the silver nitrate-adding active materials which will be loaded on the surface of the produced AgNPs. Poly(acrylonitrile-co-methyl methacrylate) copolymer (poly (AN-co-MMA)) was grafted with the prepared AgNPs. The poly(AN-co-MMA)/AgNPs were examined against ten different pathogenic bacterial strains, and the result was compared with another four different generic antibiotics. The produced poly(AN-co-MMA)/AgNPs showed high antibacterial activity compared with the four standard antibiotics. Moreover, the grafting of these AgNPs into the copolymer has potential application in the biomedical field.
Product Description
Cefazolin, European Pharmacopoeia (EP) Reference Standard
Cefotaxime acid, European Pharmacopoeia (EP) Reference Standard
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard