MilliporeSigma
  • Antitumor agents--CLXXV. Anti-tubulin action of (+)-thiocolchicine prepared by partial synthesis.
Antitumor agents--CLXXV. Anti-tubulin action of (+)-thiocolchicine prepared by partial synthesis.
Bioorganic & medicinal chemistry (1998-02-12)
Q Shi, P Verdier-Pinard, A Brossi, E Hamel, K H Lee
ABSTRACT
(+)-Thiocolchicine (2b) was prepared from (+/-)-colchicine (1) in a five-step reaction sequence that included chromatographic separation of appropriate camphanylated diastereomers. Acid hydrolysis of the (+)-diastereomer, followed by acetylation, yielded the desired product 2b. (+)-Thiocolchicine has 15-fold lower inhibitory activity against tubulin polymerization than (-)-thiocolchicine, and is 29-fold less potent for inhibiting growth of human Burkitt lymphoma cells. The enantiomer 2a, prepared from the (-)-camphanylated diastereomer, had potent activity in all assays comparable to that of (-)-thiocolchicine prepared by other methods. These results support the hypothesis that the proper configuration of colchicine-related compounds is an important requirement for their anti-tubulin action.
Sigma-Aldrich
Thiocolchicine, ≥97% (HPLC)