MilliporeSigma
  • Fulminant hepatic failure with virtually undetectable serum alkaline phosphatase.
Fulminant hepatic failure with virtually undetectable serum alkaline phosphatase.
Clinical laboratory (2014-04-05)
Gulvahid Shaikh, Mohammed Shareef, Sayed Rahman, Paul Mustacchia, Sofia Rubinstein, Steven Walerstein
ABSTRACT
Wilson's disease presenting as fulminant hepatic failure is a rare presentation that carries a high morbidity and mortality. We report a young patient who developed fulminant hepatic failure as the initial manifestation of Wilson's disease. Virtually undetectable serum alkaline phosphatase provided the first clue to the diagnosis. Our patient underwent a successful liver transplantation which is the only effective treatment in patients with Wilsonian fulminant hepatic failure. In this report, we discuss laboratory clues to the diagnosis of this form of Wilson's disease. Clinicians should have a high suspicion of Wilson's disease as any delay in diagnosis can be catastrophic.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host