MilliporeSigma
Synthesis of a library of tricyclic azepinoisoindolinones.
Beilstein journal of organic chemistry (2012-09-29)
Bettina Miller, Shuli Mao, Kara M George Rosenker, Joshua G Pierce, Peter Wipf
ABSTRACT
Hydrozirconation of 1-hexyne, the addition to in situ prepared N-acyliminium species, and ring-closing metathesis (RCM) were key steps in the preparation of a tricyclic isoindolinone scaffold. An unusual alkene isomerization process during the RCM was identified and studied in some detail. Chemical diversification for library synthesis was achieved by a subsequent alkene epoxidation and zinc-mediated aminolysis reaction. The resulting library products provided selective hits among a large number of high-throughput screens reported in PubChem, thus illustrating the utility of the novel scaffold.
Sigma-Aldrich
1-Hexyne, 97%