MilliporeSigma
  • Fabrication of non-biofouling polyethylene glycol micro- and nanochannels by ultraviolet-assisted irreversible sealing.
Fabrication of non-biofouling polyethylene glycol micro- and nanochannels by ultraviolet-assisted irreversible sealing.
Lab on a chip (2006-10-27)
Pilnam Kim, Hoon Eui Jeong, Ali Khademhosseini, Kahp Y Suh
ABSTRACT
We present a simple and widely applicable method to fabricate micro- and nanochannels comprised entirely of crosslinked polyethylene glycol (PEG) by using UV-assisted irreversible sealing to bond partially crosslinked PEG surfaces. The method developed here can be used to form channels as small as approximately 50 nm in diameter without using a sophisticated experimental setup. The manufactured channel is a homogeneous conduit made completely from non-biofouling PEG, exhibits robust sealing with minimal swelling and can be used without additional surface modification chemistries, thus significantly enhancing reliability and durability of microfluidic devices. Furthermore, we demonstrate simple analytical assays using PEG microchannels combined with patterned arrays of supported lipid bilayers (SLBs) to detect ligand (biotin)-receptor (streptavidin) interactions.
Sigma-Aldrich
Poly(ethylene glycol) diacrylate, average Mn 20,000, contains ≤1000 ppm MEHQ as inhibitor
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 μL
In Stock
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1151
$179.00
100 μL
In Stock
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1151
$515.00
0.2 mL
In Stock
Details...
A recombinant, preservative-free antibody is available for your target. Try ZRB1151
$678.00
$555.96