MilliporeSigma
  • The interplay of AMP-activated protein kinase and androgen receptor in prostate cancer cells.
The interplay of AMP-activated protein kinase and androgen receptor in prostate cancer cells.
Journal of cellular physiology (2013-10-17)
Min Shen, Zhen Zhang, Manohar Ratnam, Q Ping Dou
ABSTRACT
AMP-activated protein kinase (AMPK) has recently emerged as a potential target for cancer therapy due to the observation that activation of AMPK inhibits tumor cell growth. It is well-known that androgen receptor (AR) signaling is a major driver for the development and progression of prostate cancer and that downregulation of AR is a critical step in the induction of apoptosis in prostate cancer cells. However, little is known about the potential interaction between AMPK and AR signaling pathways. In the current study, we showed that activation of AMPK by metformin caused decrease of AR protein level through suppression of AR mRNA expression and promotion of AR protein degradation, demonstrating that AMPK activation is upstream of AR downregulation. We also showed that inhibition of AR function by an anti-androgen or its siRNA enhanced AMPK activation and growth inhibition whereas overexpression of AR delayed AMPK activation and increased prostate cancer cellular resistance to metformin treatment, suggesting that AR suppresses AMPK signaling-mediated growth inhibition in a feedback mechanism. Our findings thus reveal a novel AMPK-AR regulatory loop in prostate cancer cells and should have a potential clinical significance.
Product Description
Bicalutamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bicalutamide (CDX), ≥98% (HPLC), powder
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard