Merck
  • Deregulated balance of omega-6 and omega-3 polyunsaturated fatty acids following infection by the zoonotic pathogen Streptococcus suis.
Deregulated balance of omega-6 and omega-3 polyunsaturated fatty acids following infection by the zoonotic pathogen Streptococcus suis.
Infection and immunity (2014-02-20)
Claude Lachance, Mariela Segura, Maria C Dominguez-Punaro, Gabriella Wojewodka, Juan B De Sanctis, Danuta Radzioch, Marcelo Gottschalk
ABSTRACT
Streptococcus suis is an important swine pathogen and an emergent zoonotic pathogen. Excessive inflammation caused by S. suis is responsible for early high mortality in septic shock-like syndrome cases. Polyunsaturated fatty acids (PUFAs) may contribute to regulating inflammatory processes. This study shows that mouse infection by S. suis is accompanied by an increase of arachidonic acid, a proinflammatory omega-6 (ω-6) PUFA, and by a decrease of docosahexaenoic acid, an anti-inflammatory ω-3 PUFA. Macrophages infected with S. suis showed activation of mitogen-activated protein kinase pathways and cyclooxygenase-2 upregulation. Fenretinide, a synthetic vitamin A analog, reduced in vitro expression of inflammatory mediators. Pretreatment of mice with fenretinide significantly improved their survival by reducing systemic proinflammatory cytokines during the acute phase of an S. suis infection. These findings indicate a beneficial effect of fenretinide in diminishing the expression of inflammation and improving survival during an acute infection by a virulent S. suis strain.
Sigma-Aldrich
Retinoic acid p-hydroxyanilide, ≥95%