MilliporeSigma
  • Amino-acid mutations to extend the biological half-life of a therapeutically valuable mutant of human butyrylcholinesterase.
Amino-acid mutations to extend the biological half-life of a therapeutically valuable mutant of human butyrylcholinesterase.
Chemico-biological interactions (2014-03-04)
Lei Fang, Shurong Hou, Liu Xue, Fang Zheng, Chang-Guo Zhan
ABSTRACT
Cocaine is a widely abused and addictive drug without an FDA-approved medication. Our recently designed and discovered cocaine hydrolase, particularly E12-7 engineered from human butyrylcholinesterase (BChE), has the promise of becoming a valuable cocaine abuse treatment. An ideal anti-cocaine therapeutic enzyme should have not only a high catalytic efficiency against cocaine, but also a sufficiently long biological half-life. However, recombinant human BChE and the known BChE mutants have a much shorter biological half-life compared to the native human BChE. The present study aimed to extend the biological half-life of the cocaine hydrolase without changing its high catalytic activity against cocaine. Our strategy was to design possible amino-acid mutations that can introduce cross-subunit disulfide bond(s) and, thus, change the distribution of the oligomeric forms and extend the biological half-life. Three new BChE mutants (E364-532, E377-516, and E535) were predicted to have a more stable dimer structure with the desirable cross-subunit disulfide bond(s) and, therefore, a different distribution of the oligomeric forms and a prolonged biological half-life. The rational design was followed by experimental tests in vitro and in vivo, confirming that the rationally designed new BChE mutants, i.e. E364-532, E377-516, and E535, indeed had a remarkably different distribution of the oligomeric forms and prolonged biological half-life in rats from ∼7 to ∼13h without significantly changing the catalytic activity against (-)-cocaine. This is the first demonstration that rationally designed amino-acid mutations can significantly prolong the biological half-life of a high-activity enzyme without significantly changing the catalytic activity.
Product Description
Supelco
Cocaine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cocaine hydrochloride solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
Cocaine hydrochloride
Sigma-Aldrich
Cocaine free base