MilliporeSigma
  • Hybrid macroporous gelatin/bioactive-glass/nanosilver scaffolds with controlled degradation behavior and antimicrobial activity for bone tissue engineering.
Hybrid macroporous gelatin/bioactive-glass/nanosilver scaffolds with controlled degradation behavior and antimicrobial activity for bone tissue engineering.
Journal of biomedical nanotechnology (2014-04-23)
M Yazdimamaghani, D Vashaee, S Assefa, K J Walker, S V Madihally, G A Köhler, L Tayebi
ABSTRACT
A new composition of gelatin/bioactive-glass/silver nanoparticle was synthesized and employed to prepare antibacterial macroporous scaffolds with potential applications in bone tissue engineering. A set of macroporous nanocomposite scaffolds were developed from an aqueous solution of gelatin by freeze-drying and crosslinking using genipin at ambient temperature. Silver nanoparticles were successfully synthesized in situ in gelatin solution by heat treatment reduction as a simple and "green" method in which gelatin acted as a natural reducing and stabilizing agent. The effect of the incorporation of the bioactive-glass and the silver nanoparticle concentration on the physicochemical properties of the scaffolds, such as the gel fraction, porosity, in vitro enzyme degradation, morphology, and swelling behavior was studied. Furthermore, the in vitro viability of human mesenchymal stem cells (hMSC) and the antibacterial activity against gram-negative Escherichia coli and gram-positive Staphylococcus aureus were tested on the scaffolds. It was found that upon the addition of silver nanoparticles the porosity, pore size, swelling, and antibacterial properties were enhanced. The silver nanoparticles increased the in vitro enzyme degradation in samples without bioactive-glass; however, the degradation was remarkably reduced by addition of bioactive-glass. In addition, formation of apatite particles, the main inorganic constituent of the bone, on the surface of the bioactive-glass containing scaffolds were confirmed after immersion in simulated body fluid (SBF). The viability of hMSC on the scaffold suggested that gelatin/bioactive-glass/nanosilver scaffolds can be used as an antibacterial scaffolds.
Product Description
Silver, foil, 15mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.0025mm, 99.97%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silver, foil, 6mm disks, thickness 0.015mm, 99.97%
Silver, foil, 5m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 6mm disks, thickness 0.008mm, 99.97%
Silver, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.007mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, 8mm disks, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 6mm disks, thickness 0.5mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.006mm, 99.97%
Silver, foil, 4mm disks, thickness 0.02mm, 99.97%
Silver, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Silver, foil, 15mm disks, thickness 0.015mm, 99.97%
Silver, foil, 15mm disks, thickness 0.008mm, 99.97%
Silver, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Silver, foil, 25mm disks, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95+%
Silver, foil, 50mm disks, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 4mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 4mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 50mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.20mm, as rolled, 99.95+%