MilliporeSigma
  • ATSDR evaluation of health effects of chemicals. VI. Di(2-ethylhexyl)phthalate. Agency for Toxic Substances and Disease Registry.
ATSDR evaluation of health effects of chemicals. VI. Di(2-ethylhexyl)phthalate. Agency for Toxic Substances and Disease Registry.
Toxicology and industrial health (2000-04-29)
M Fay, J M Donohue, C De Rosa
ABSTRACT
Di(2-ethylhexyl)phthalate (also known as DEHP, bis(2-ethylhexyl)phthalate, or BEHP; CAS Registry Number 117-81-7) is a widely-used plasticizer. It is found in numerous plastic articles, such as paints, inks, floor tiles, upholstery, shower curtains, footwear, plastic bags, food-packaging materials, toys, and medical tubing. Not surprisingly, DEHP appears at many waste sites. As part of its mandate, the Agency for Toxic Substances and Disease Registry (ATSDR) prepares toxicological profiles on hazardous chemicals that are of greatest public health concern at Comprehensive Environmental Response, Compensation, and Liability Act (CERCLA) National Priority List (NPL) sites. These profiles comprehensively summarize toxicological and environmental information. This article constitutes the release of the bulk of ATSDR's profile for DEHP (ATSDR, 1993) into the mainstream scientific literature. An extensive listing of human and animal health effects, organized by route, duration, and endpoint, is presented. Toxicological information on toxicokinetics, biomarkers, interactions, sensitive subpopulations, reducing toxicity after exposure, and relevance to public health is also included. Environmental information encompasses physical properties, production and use, environmental fate, levels seen in the environment, analytical methods, and a listing of regulations. ATSDR, at the behest of Congress and therefore the citizenry, prepares these profiles to inform the public about site contaminants.
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol