MilliporeSigma
  • Review of the extrinsic stain removal and enamel/dentine abrasion by a calcium carbonate and perlite containing whitening toothpaste.
Review of the extrinsic stain removal and enamel/dentine abrasion by a calcium carbonate and perlite containing whitening toothpaste.
International dental journal (2006-09-16)
Andrew Joiner
ABSTRACT
There has been an increase in the demand from consumers and patients for products that whiten teeth. To meet this demand, a whitening toothpaste containing calcium carbonate and perlite as the abrasive system and an efficacious fluoride source has recently been launched. The aim of the current paper is to review the toothpaste's stain removal efficacy and its effects on enamel and dentine wear. It has been shown to be effective at removing model extrinsic stain in vitro. Further, it has been shown to be more effective in removing naturally occurring extrinsic tooth stain than a silica non-whitening control toothpaste after two weeks of twice daily brushing in a parallel group, double-blind clinical study using 152 adult volunteers. In addition, the enhanced whitening effect did not give a clinically relevant level of wear to enamel or a significant increase in dentine wear compared to marketed non-whitening toothpaste formulations, as shown by using an in situ type model with ex vivo brushing.
Sigma-Aldrich
Calcium carbonate, CP
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Marble, CP
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard