MilliporeSigma
  • The use of fillers and botulinum toxin type A in combination with superficial glycolic acid (alpha-hydroxy acid) peels: optimizing injection therapy with the skin-smoothing properties of peels.
The use of fillers and botulinum toxin type A in combination with superficial glycolic acid (alpha-hydroxy acid) peels: optimizing injection therapy with the skin-smoothing properties of peels.
Cutis (2007-04-26)
Marta I Rendon, Cheryl Effron, Brenda L Edison
ABSTRACT
There are many procedures that a physician may utilize to improve the appearance and quality of the skin. Combining procedures can enhance the overall result and lead to increased patient satisfaction. Thus, it is important to choose procedures that will complement each other. Fillers or botulinum toxin type A (BTX-A) can plump the skin and smooth lines and wrinkles but will do little for uneven tone, skin laxity, or radiance and clarity. These signs of aging can be addressed with superficial glycolic acid peels. Methods of combining injectable compounds with superficial glycolic acid peels were discussed at a dermatologist roundtable event and are summarized in this article.
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Glycolic acid solution, CP, 70% in H2O
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycolic acid, SAJ special grade, ≥98.0%
Supelco
Glycolic Acid, Pharmaceutical Secondary Standard; Certified Reference Material