MilliporeSigma
Fragrance material review on nerolidol (isomer unspecified).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of nerolidol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans