MilliporeSigma
The role of zinc and copper in autism spectrum disorders.
Acta neurobiologiae experimentalis (2013-07-05)
Geir Bjorklund
ABSTRACT
Autism spectrum disorders (ASDs) are a group of developmental disabilities that can cause significant social, communication and behavioral challenges. Several studies have suggested a disturbance in the copper (Cu) and zinc (Zn) metabolism in ASDs. Zinc deficiency, excess Cu levels, and low Zn/Cu ratio are common in children diagnosed with an ASD. The literature also suggests that mercury accumulation may occur as a cause or consequence of metallothionein (MT) dysfunction in children diagnosed with an ASD, which may be one of the causes of Zn deficiency. MTs are proteins with important functions in metal metabolism and protection. Zinc and Cu bind to and participate in the control of the synthesis of MT proteins. Studies indicate that the GABAergic system may be involved in ASDs, and that Zn and Cu may play a role in this system.
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Copper, wire reel, 100m, diameter 1.0mm, annealed, 99.9%
Copper, wire reel, 10m, diameter 1.0mm, annealed, 99.9%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.025mm, hard, 99.95+%
Copper, wire reel, 0.2m, diameter 0.1mm, aluminum cladding, outside diameter 0.25mm, 99.9%
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper, wire reel, 2000m, diameter 0.075mm, annealed, 99.9%
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Copper, wire reel, 2m, diameter 0.75mm, as drawn, 99.98+%
Copper, wire reel, 500m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Copper, wire reel, 50m, diameter 0.25mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%