- Ocular manifestations of monoclonal copper-binding immunoglobulin.
Ocular manifestations of monoclonal copper-binding immunoglobulin.
Survey of ophthalmology (2013-10-12)
Sejal Shah, Edgar M Espana, Curtis E Margo
PMID24112547
ABSTRACT
The dense accumulation of copper in Descemet membrane and lens capsule is the characteristic manifestation of a circulating monoclonal antibody with strong affinity for copper. The overproduction of this monoclonal immunoglobulin may be associated with either multiple myeloma or a benign monoclonal gammopathy. Despite prolonged exposure to elevated serum copper, no other tissues in the body are adversely affected by this redox metal. We describe the clinical and pathological findings in a 46-year-old woman with this disorder.
Product Description
Continuous cast copper (O), BCR®, certified reference material, rod
Copper - O.F.H.C., foil, 25mm disks, thickness 0.025mm, hard, 99.95+%
Copper, wire reel, 0.2m, diameter 0.1mm, aluminum cladding, outside diameter 0.25mm, 99.9%
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper, wire reel, 2000m, diameter 0.075mm, annealed, 99.9%
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Copper, wire reel, 2m, diameter 0.75mm, as drawn, 99.98+%
Copper, wire reel, 500m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Copper, wire reel, 50m, diameter 0.25mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 12.7mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 100x100mm, thickness 0.1mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.025mm, hard, 99.95+%