MilliporeSigma
  • Bacterial sodium channels: models for eukaryotic sodium and calcium channels.
Bacterial sodium channels: models for eukaryotic sodium and calcium channels.
Handbook of experimental pharmacology (2014-04-17)
Todd Scheuer
ABSTRACT
Eukaryotic sodium and calcium channels are made up of four linked homologous but different transmembrane domains. Bacteria express sodium channels comprised of four identical subunits, each being analogous to a single homologous domain of their eukaryotic counterparts. Key elements of primary structure are conserved between bacterial and eukaryotic sodium and calcium channels. The simple protein structure of the bacterial channels has allowed extensive structure-function probes of key regions as well as allowing determination of several X-ray crystallographic structures of these channels. The structures have revealed novel features of sodium and calcium channel pores and elucidated the structural importance of many of the conserved features of primary sequence. The structural information has also formed the basis for computational studies probing the basis for sodium and calcium selectivity and gating.
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sodium, CP
Sigma-Aldrich
Calcium, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)