Merck
  • Cefazolin-induced pseudomembranous colitis resulting in perforation of the sigmoid colon.
Cefazolin-induced pseudomembranous colitis resulting in perforation of the sigmoid colon.
Diseases of the colon and rectum (1986-02-01)
T Osler, D Lott, J Bordley, F Lynch, C Ellsworth, A Kozak
ABSTRACT
The seventh case of probable cefazolin-induced pseudomembranous colitis is reported. Perforation of the colon necessitated sigmoid resection. The postoperative course was protracted, and illustrates the difficulty of managing advanced pseudomembranous colitis when the oral route of antibiotic administration is not available. Although rare, pseudomembranous colitis related to cefazolin administration is a potentially fatal complication. The routine use of prophylactic antibiotics must be weighed against this possibility.
Product Description
Cefazolin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Cefazolin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.