MilliporeSigma
Stearic acid metabolism and atherogenesis: history.
The American journal of clinical nutrition (1994-12-01)
D Kritchevsky
ABSTRACT
Studies conducted in dogs, rats, and hamsters show that stearic acid or stearic acid-rich glycerides are absorbed less efficiently than are lauric, myristic, and palmitic acids or their triglycerides. This observation may explain in part why stearic acid is less cholesterolemic than saturated fatty acids of shorter chain length. In rabbits, cocoa butter or other fats rich in stearic acid are less atherogenic than other saturated fatty acids. This finding is true for both cholesterol-containing and cholesterol-free diets.
Product Description
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, reagent grade, 95%