Merck
2-Alkenal-scavenging ability of m-diphenols.
Food chemistry (2014-05-07)
Francisco J Hidalgo, Rosario Zamora
ABSTRACT
The reaction between m-diphenols (resorcinol, 2-methylresorcinol, 2,5-dimethylresorcinol, 3-methylphenol, orcinol, and phloroglucinol) and 2-alkenals (2-pentenal and 2-octenal) was studied in an attempt to understand the chemical pathways involved in the scavenging ability of m-diphenols for the 2-alkenals produced as a consequence of lipid oxidation. Phenols reacted chemically with 2-alkenals producing a number of 2H-chromenols, chromandiols, chromanols, and dihydropyrano[3,2-g]chromenes, which were isolated and identified by 1D and 2D nuclear magnetic resonance (NMR) spectroscopy and mass spectrometry (MS). The identification of all these compounds resulted in the construction of a general pathway for these reactions. These results confirm that the 2-alkenal-scavenging ability of m-diphenols is a consequence of their structure. This is a complex reaction in which many different products are formed. The most stable products were the chromandiols. However, the main reaction products were the 2H-chromenols. These products were unstable and disappeared as a consequence of polymerisation and browning reactions.
Sigma-Aldrich
2-Methylresorcinol, 98%