Merck
  • Optically active cyclohexene derivative as a new antisepsis agent: an efficient synthesis of ethyl (6R)-6-[N-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate (TAK-242).
Optically active cyclohexene derivative as a new antisepsis agent: an efficient synthesis of ethyl (6R)-6-[N-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate (TAK-242).
Chemical & pharmaceutical bulletin (2006-01-06)
Masami Yamada, Takashi Ichikawa, Toru Yamano, Fumio Kikumoto, Yuji Nishikimi, Norikazu Tamura, Tomoyuki Kitazaki
ABSTRACT
Two new synthetic methods were established for the efficient synthesis of optically active cyclohexene antisepsis agent, ethyl (6R)-6-[N-(2-chloro-4-fluorophenyl)sulfamoyl]cyclohex-1-ene-1-carboxylate [(R)-1: TAK-242)]. The first method involved recrystallization from methanol of the diastereomeric mixture (6RS,1'R)-7, obtained by esterification of carboxylic acid 3 with (S)-1-(4-nitrophenyl)ethanol [(S)-5)] to give the desired isomer (6R,1'R)-7 with 99% de in 32% yield. Subsequent catalytic hydrogenolysis and esterification gave (R)-1 with >99% ee. The second method employed enantioselective hydrolysis of acetoxymethyl ester 9a (prepared by alkylation of 3 with bromomethyl acetate) with Lipase PS-D to give the eutomeric enantiomer (R)-9a with excellent enantioselectivity (>99% ee) and high yield (48%). The desired (R)-1 was then obtained by transesterification with ethanol in the presence of concentrated sulfuric acid without loss of ee. Of these, the procedure employing enzymatic kinetic resolution using Lipase PS-D is the more efficient and practical preparation of (R)-1.
Sigma-Aldrich
Bromomethyl acetate, 95%