- Ferrisiderophore reductase activity in Agrobacterium tumefaciens.
Ferrisiderophore reductase activity in Agrobacterium tumefaciens.
Journal of bacteriology (1982-02-01)
J S Lodge, C G Gaines, J E Arceneaux, B R Byers
PMID7056702
ABSTRACT
Reduction of the iron in ferriagrobactin by the cytoplasmic fraction of Agrobacterium tumefaciens strictly required NaDH as the reductant. Addition of flavin mononucleotide and anaerobic conditions were necessary for the reaction; when added with flavin mononucleotide, magnesium was stimulatory. This ferrisiderophore reductase activity may be a part of the iron assimilation process in A. tumefaciens.
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder