MilliporeSigma
  • PPARγ and RXR ligands disrupt the inflammatory cross-talk in the hypoxic breast cancer stem cells niche.
PPARγ and RXR ligands disrupt the inflammatory cross-talk in the hypoxic breast cancer stem cells niche.
Journal of cellular physiology (2014-03-08)
Alessio Papi, Sabrina De Carolis, Sara Bertoni, Gianluca Storci, Virginia Sceberras, Donatella Santini, Claudio Ceccarelli, Mario Taffurelli, Marina Orlandi, Massimiliano Bonafé
ABSTRACT
Cancer stem cells (CSCs) are affected by the local micro-environment, the niche, in which inflammatory stimuli and hypoxia act as steering factors. Here, two nuclear receptors (NRs) agonists, i.e. pioglitazone (PGZ), a ligand of peroxisome proliferator activated receptor-γ, and 6-OH-11-O-hydroxyphenanthrene (IIF), a ligand of retinoid X receptors, were investigated for their capability to interference with the cross-talk between breast CSCs and the niche compartment. We found that IIF potentiates the ability of PGZ to hamper the mammospheres-forming capability of human breast tumours and MCF7 cancer cells, reducing the expression of CSCs regulatory genes (Notch3, Jagged1, SLUG, Interleukin-6, Apolipoprotein E, Hypoxia inducible factor-1α and Carbonic anhydrase IX). Notably, these effects are not observed in normal-MS obtained from human breast tissue. Importantly, NRs agonists abolish the capability of hypoxic MCF7 derived exosomes to induce a pro-inflammatory phenotype in mammary glands fibroblasts. Moreover, NRs agonist also directly acts on breast tumour associated fibroblasts to downregulate nuclear factor-κB pathway and metalloproteinases (MMP2 and MMP9) expression and activity. In conclusion, NRs agonists disrupt the inflammatory cross-talk of the hypoxic breast CSCs niche.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Pioglitazone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
USP
Pioglitazone hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Sigma-Aldrich
Pioglitazone hydrochloride, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490
Pioglitazone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
In Stock
Details...
₩57,246
250 g
Estimated to ship on April 14, 2025
Details...
₩116,725
1 kg
Estimated to ship on April 14, 2025
Details...
₩283,490