MilliporeSigma
  • Variability in urinary phthalate metabolite levels across pregnancy and sensitive windows of exposure for the risk of preterm birth.
Variability in urinary phthalate metabolite levels across pregnancy and sensitive windows of exposure for the risk of preterm birth.
Environment international (2014-06-18)
Kelly K Ferguson, Thomas F McElrath, Yi-An Ko, Bhramar Mukherjee, John D Meeker
ABSTRACT
Preterm birth is a significant public health problem, affecting over 1 in 10 live births and contributing largely to infant mortality and morbidity. Everyday exposure to environmental chemicals such as phthalates could contribute to prematurity, and may be modifiable. In the present study we examine variability in phthalate exposure across gestation and identify windows of susceptibility for the relationship with preterm birth. Women were recruited early in pregnancy as part of a prospective, longitudinal birth cohort at the Brigham and Women's Hospital in Boston, Massachusetts. Urine samples were collected at up to 4 time points during gestation for phthalate measurement, and birth outcomes were recorded at delivery. From this population we selected all 130 cases of preterm birth, defined as delivery before 37 weeks of completed gestation, as well as 352 random controls. Urinary phthalate metabolite levels were moderately variable over pregnancy, but levels measured at multiple time points were associated with increased odds of preterm birth. Adjusted odds ratios (aOR) for spontaneous preterm birth were strongest in association with phthalate metabolite concentrations measured at the beginning of the third trimester (aOR for summed di-2-ethylhexyl phthalate metabolites [∑DEHP]=1.33, 95% confidence interval [CI]=1.02, 1.73). Odds ratios for placental preterm birth, defined as delivery with presentation of preeclampsia or intrauterine growth restriction, were slightly elevated in the first trimester for DEHP metabolites (aOR for ∑DEHP=1.33, 95% CI=0.99, 1.78). Pregnant women with exposure to phthalates both early and late in pregnancy are at an increased risk of delivering preterm, but mechanisms may differ based on etiology.
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Phthalic acid, SAJ special grade, ≥99.0%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and Inventory is not available. Please check the Millipore website for more information.
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Phthalic acid, analytical standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Potassium phthalate monobasic, JIS special grade, 99.8-100.2%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Pricing and Inventory is not available. Please check the Millipore website for more information.
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Pricing and Inventory is not available. Please check the Millipore website for more information.
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Pricing and Inventory is not available. Please check the Millipore website for more information.