MilliporeSigma
  • The enigmatic plasmacytoid T cells develop into dendritic cells with interleukin (IL)-3 and CD40-ligand.
The enigmatic plasmacytoid T cells develop into dendritic cells with interleukin (IL)-3 and CD40-ligand.
The Journal of experimental medicine (1997-03-17)
G Grouard, M C Rissoan, L Filgueira, I Durand, J Banchereau, Y J Liu
ABSTRACT
A subset of CD4+CD11c-CD3- blood cells was recently shown to develop into dendritic cells when cultured with monocyte conditioned medium. Here, we demonstrate that CD4+ CD11c-CD3- cells, isolated from tonsils, correspond to the so-called plasmacytoid T cells, an obscure cell type that has long been observed by pathologists within secondary lymphoid tissues. They express CD45RA, but not markers specific for known lymphoid- or myeloid-derived cell types. They undergo rapid apoptosis in culture, unless rescued by IL-3. Further addition of CD40-ligand results in their differentiation into dendritic cells that express low levels of myeloid antigens CD13 and CD33.
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution